missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiophene, 98+%

Catalog No. AAA1339422
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1339422 100 g
AAA1339436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1339422 Supplier Thermo Scientific Chemicals Supplier No. A1339422

CAS: 616-44-4 | C5H6S | 98.163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 616-44-4
Molecular Formula C5H6S
Molecular Weight (g/mol) 98.163
MDL Number MFCD00005470
InChI Key QENGPZGAWFQWCZ-UHFFFAOYSA-N
Synonym thiophene, 3-methyl, 3-thiotolene, beta-methylthiophene, poly, p3mt, 3-methyl thiophene, 3-methyl-thiophene, methyl-3-thiophene, ccris 2937, thiophene, 3-methyl-, homopolymer
PubChem CID 12024
IUPAC Name 3-methylthiophene
SMILES CC1=CSC=C1

Specifications

Melting Point -69°C
Density 1.02
Boiling Point 115°C to 117°C
Flash Point 11°C (51°F)
Odor Aromatic
Refractive Index 1.52
Quantity 100 g
UN Number UN1993
Beilstein 1300
Formula Weight 98.17
Percent Purity ≥98%
Chemical Name or Material 3-Methylthiophene
Show More Show Less
Hazard Category H225-H302-H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H225-H332
Highly flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 210-482-6
RTECSNumber XM9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only