missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiophene-2-methylamine, 96%

Catalog No. p-7047226
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL18094MD 250 mg
AAL1809403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL18094MD Supplier Thermo Scientific Chemicals Supplier No. L18094MD
Only null left

CAS: 104163-35-1 | C6H9NS | 127.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104163-35-1
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.205
MDL Number MFCD02093988
InChI Key SWZNXCABBUKIPZ-UHFFFAOYSA-N
PubChem CID 2798756
IUPAC Name (3-methylthiophen-2-yl)methanamine
SMILES CC1=C(SC=C1)CN

Specifications

Boiling Point 78°C (4 mmHg)
Refractive Index 1.5585
Quantity 250 mg
UN Number UN2735
Beilstein 108864
Sensitivity Air Sensitive
Synonym 3-methylthiophen-2-yl methanamine, 3-methyl-2-thienyl methylamine, 3-methylthiophene-2-methylamine, 2-thiophenemethanamine,3-methyl, 2-thiophenemethanamine, 3-methyl, acmc-20apo3, 3-metylthiophene-2-metylamine, 2-aminomethyl-3-methylthiophene, 3-methylthiophene-2-methanamine, 3-methylthien-2-yl methylamine
Formula Weight 127.21
Percent Purity 98%
Chemical Name or Material 3-Methylthiophene-2-methylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only