missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiophene-2-carboxylic acid, 98%

Catalog No. AAA1919406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1919406 5 g
AAA1919414 25 g
AAA1919422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1919406 Supplier Thermo Scientific Chemicals Supplier No. A1919406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 23806-24-8 | C6H5O2S | 141.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23806-24-8
Molecular Formula C6H5O2S
Molecular Weight (g/mol) 141.16
MDL Number MFCD00005438
InChI Key IFLKEBSJTZGCJG-UHFFFAOYSA-M
Synonym 3-methyl-2-thiophenecarboxylic acid, 3-methyl-2-thenoic acid, 2-thiophenecarboxylic acid, 3-methyl, 2-carboxy-3-methylthiophene, 3-methyl-2-thiophene carboxylic acid, 3-methyl-2-thiophenecarboxylic, 3-methyl-thiophene-2-carboxylic acid, 3-methyl-2-thiophenecarboxylicacid, pubchem5182, acmc-1cinc
PubChem CID 90269
IUPAC Name 3-methylthiophene-2-carboxylic acid
SMILES CC1=C(SC=C1)C([O-])=O

Specifications

Melting Point 144°C to 149°C
Quantity 5 g
Beilstein 116184
Formula Weight 142.18
Percent Purity 98%
Chemical Name or Material 3-Methylthiophene-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 245-894-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only