missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Methylthio)propionic Acid 98.0+%, TCI America™
Supplier: TCI America M0811100G
Specifications
| 3-(Methylthio)propionic Acid | |
| Yellow | |
| C4H8O2S | |
| 100 g | |
| 3-methylthio propionic acid, 3-methylthiopropionate, 3-methylthiopropionic acid, 4-thiapentanoic acid, 3-methylsulfanyl propanoic acid, propanoic acid, 3-methylthio, 3-methylmercaptopropionic acid, unii-f523ali47d, 3-methylthio propanoic acid, propionic acid, 3-methylthio | |
| CSCCC(=O)O | |
| 120.166 | |
| CHEBI:1438 | |
| ≥98.0% (GC,T) |
| 646-01-5 | |
| 130°C | |
| MFCD00059635 | |
| 3265 | |
| CAOMCZAIALVUPA-UHFFFAOYSA-N | |
| 3-methylsulfanylpropanoic acid | |
| 563 | |
| 120.17 | |
| Liquid |
Chemical Identifiers
| 646-01-5 | |
| 120.166 | |
| CAOMCZAIALVUPA-UHFFFAOYSA-N | |
| 563 | |
| 3-methylsulfanylpropanoic acid |
| C4H8O2S | |
| MFCD00059635 | |
| 3-methylthio propionic acid, 3-methylthiopropionate, 3-methylthiopropionic acid, 4-thiapentanoic acid, 3-methylsulfanyl propanoic acid, propanoic acid, 3-methylthio, 3-methylmercaptopropionic acid, unii-f523ali47d, 3-methylthio propanoic acid, propionic acid, 3-methylthio | |
| CHEBI:1438 | |
| CSCCC(=O)O |
Safety and Handling
TSCA : No