missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)propionic Acid 98.0+%, TCI America™

Catalog No. M081125G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M081125G 25 g
M0811100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M081125G Supplier TCI America Supplier No. M081125G
Only null left

Chemical Identifiers

CAS 646-01-5
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.166
MDL Number MFCD00059635
InChI Key CAOMCZAIALVUPA-UHFFFAOYSA-N
Synonym 3-methylthio propionic acid, 3-methylthiopropionate, 3-methylthiopropionic acid, 4-thiapentanoic acid, 3-methylsulfanyl propanoic acid, propanoic acid, 3-methylthio, 3-methylmercaptopropionic acid, unii-f523ali47d, 3-methylthio propanoic acid, propionic acid, 3-methylthio
PubChem CID 563
ChEBI CHEBI:1438
IUPAC Name 3-methylsulfanylpropanoic acid
SMILES CSCCC(=O)O

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 g
UN Number 3265
Formula Weight 120.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Methylthio)propionic Acid