missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylsulfolane 98.0+%, TCI America™
Supplier: TCI America M0436250G
Specifications
| 3-Methylsulfolane | |
| Yellow | |
| C5H10O2S | |
| 250 g | |
| CMJLMPKFQPJDKP-UHFFFAOYSA-N | |
| 3-methylthiolane 1,1-dioxide | |
| 79119 | |
| ≥98.0% (GC) |
| 872-93-5 | |
| 104°C | |
| MFCD00005487 | |
| 3-methylsulfolane, 3-methyl sulfolane, 3-methyltetrahydrothiophene 1,1-dioxide, thiophene, tetrahydro-3-methyl-, 1,1-dioxide, acmc-209qjh, 3-methylthiolane-1,1-dione, 3-methylthiacyclopentane 1,1-dioxide, 3-methyl-2,4-dihydrothiophene 1,1-dioxide, thiophene,tetrahydro-3-methyl-, 1,1-dioxide, 1,1-dioxo-1??-thiolan-3-yl methyl | |
| CC1CCS(=O)(=O)C1 | |
| 134.193 | |
| 134.19 | |
| Liquid |
Chemical Identifiers
| 872-93-5 | |
| 134.193 | |
| CMJLMPKFQPJDKP-UHFFFAOYSA-N | |
| 79119 | |
| CC1CCS(=O)(=O)C1 |
| C5H10O2S | |
| MFCD00005487 | |
| 3-methylsulfolane, 3-methyl sulfolane, 3-methyltetrahydrothiophene 1,1-dioxide, thiophene, tetrahydro-3-methyl-, 1,1-dioxide, acmc-209qjh, 3-methylthiolane-1,1-dione, 3-methylthiacyclopentane 1,1-dioxide, 3-methyl-2,4-dihydrothiophene 1,1-dioxide, thiophene,tetrahydro-3-methyl-, 1,1-dioxide, 1,1-dioxo-1??-thiolan-3-yl methyl | |
| 3-methylthiolane 1,1-dioxide |
Safety and Handling
TSCA : Yes