missing translation for 'onlineSavingsMsg'
Learn More

3-Methylsulfolane 98.0+%, TCI America™

Catalog No. M043625G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M043625G 25 g
M0436250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M043625G Supplier TCI America Supplier No. M043625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 872-93-5
Molecular Formula C5H10O2S
Molecular Weight (g/mol) 134.193
MDL Number MFCD00005487
InChI Key CMJLMPKFQPJDKP-UHFFFAOYSA-N
Synonym 3-methylsulfolane, 3-methyl sulfolane, 3-methyltetrahydrothiophene 1,1-dioxide, thiophene, tetrahydro-3-methyl-, 1,1-dioxide, acmc-209qjh, 3-methylthiolane-1,1-dione, 3-methylthiacyclopentane 1,1-dioxide, 3-methyl-2,4-dihydrothiophene 1,1-dioxide, thiophene,tetrahydro-3-methyl-, 1,1-dioxide, 1,1-dioxo-1??-thiolan-3-yl methyl
PubChem CID 79119
IUPAC Name 3-methylthiolane 1,1-dioxide
SMILES CC1CCS(=O)(=O)C1

Specifications

Color Yellow
Boiling Point 104°C
Quantity 25 g
Formula Weight 134.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylsulfolane