missing translation for 'onlineSavingsMsg'
Learn More

3-Methylsalicylaldehyde 98.0+%, TCI America™

Catalog No. p-7133126
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22705G 5 g
M227025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22705G Supplier TCI America Supplier No. M22705G

Chemical Identifiers

CAS 824-42-0
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00040798
InChI Key IPPQNXSAJZOTJZ-UHFFFAOYSA-N
Synonym 3-methylsalicylaldehyde, 2,3-cresotaldehyde, 2-hydroxy-3-methyl-benzaldehyde, 3-methylsalicylic aldehyde, 3-methyl-2-hydroxybenzaldehyde, benzaldehyde, hydroxy-3-methyl, benzaldehyde, 2-hydroxy-3-methyl, zlchem 747, pubchem14264, 2-formyl-6-methylphenol
PubChem CID 522777
ChEBI CHEBI:20110
IUPAC Name 2-hydroxy-3-methylbenzaldehyde
SMILES CC1=CC=CC(=C1O)C=O

Specifications

Melting Point 17°C
Color Yellow
Boiling Point 209°C
Quantity 5 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Methylsalicylaldehyde