missing translation for 'onlineSavingsMsg'
Learn More

3-Methylquinoline 98.0+%, TCI America™

Catalog No. M078225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M078225G 25 g
M0782500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M078225G Supplier TCI America Supplier No. M078225G

Chemical Identifiers

CAS 612-58-8
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00014661
InChI Key DTBDAFLSBDGPEA-UHFFFAOYSA-N
Synonym quinoline, 3-methyl, 3-methyl-1-benzazine, 3-methyl-quinoline, ccris 2896, unii-1yc9rbw2g0, beta-methylquinoline, 1yc9rbw2g0, 3-methyl quinoline, pubchem2344, acmc-209mql
PubChem CID 11926
ChEBI CHEBI:20140
IUPAC Name 3-methylquinoline
SMILES CC1=CN=C2C=CC=CC2=C1

Specifications

Color Yellow
Boiling Point 255°C
Quantity 25 g
Formula Weight 143.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylquinoline
EINECSNumber (5)-0796
RTECSNumber VC0527000
TSCA No