missing translation for 'onlineSavingsMsg'
Learn More

3-Methylquinoline, 98+%

Catalog No. AAA1310906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1310906 5 g
AAA1310914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1310906 Supplier Thermo Scientific Chemicals Supplier No. A1310906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 612-58-8 | C10H9N | 143.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 612-58-8
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00014661
InChI Key DTBDAFLSBDGPEA-UHFFFAOYSA-N
Synonym quinoline, 3-methyl, 3-methyl-1-benzazine, 3-methyl-quinoline, ccris 2896, unii-1yc9rbw2g0, beta-methylquinoline, 1yc9rbw2g0, 3-methyl quinoline, pubchem2344, acmc-209mql
PubChem CID 11926
ChEBI CHEBI:20140
IUPAC Name 3-methylquinoline
SMILES CC1=CN=C2C=CC=CC2=C1

Specifications

Melting Point 15°C
Density 1.069
Boiling Point 252°C to 253°C
Flash Point >110°C (230°F)
Refractive Index 1.615
Quantity 5 g
Beilstein 110325
Formula Weight 143.19
Percent Purity ≥98%
Chemical Name or Material 3-Methylquinoline
Hazard Category H302-H315-H319-H335-H341
Hazard Statement GHS H Statement
H341-H315-H319
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
EINECSNumber 210-315-7
RTECSNumber VC0527000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only