missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpyridine 98.0+%, TCI America™

Catalog No. P041625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P041625ML 25 mL
P0416500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P041625ML Supplier TCI America Supplier No. P041625ML

Chemical Identifiers

CAS 108-99-6
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.129
MDL Number MFCD00006402
InChI Key ITQTTZVARXURQS-UHFFFAOYSA-N
Synonym 3-picoline, beta-picoline, pyridine, 3-methyl, m-picoline, m-methylpyridine, b-picoline, beta-methylpyridine, .beta.-methylpyridine, .beta.-picoline, meta-methylpyridine
PubChem CID 7970
ChEBI CHEBI:39922
IUPAC Name 3-methylpyridine
SMILES CC1=CN=CC=C1

Specifications

Melting Point -18°C
Color Yellow
Boiling Point 142°C
Quantity 25 mL
UN Number 2313
Formula Weight 93.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylpyridine
EINECSNumber (5)-0711
RTECSNumber TJ5000000
TSCA Yes