missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylpyrazole 97.0+%, TCI America™
Supplier: TCI America M056425G
Specifications
| 3-Methylpyrazole | |
| Yellow | |
| C4H6N2 | |
| 25 g | |
| 3-methylpyrazole, 3-methyl-1h-pyrazole, 1h-pyrazole, 3-methyl, 5-methylpyrazole, 3 5-methylpyrazole, pyrazole, 5-methyl, 1h-pyrazole,5-methyl, 1h-pyrazole, 5-methyl, pyrazole, 3-methyl, unii-w7ku1rro6u | |
| CC1=CC=NN1 | |
| 82.11 | |
| 82.11 | |
| Liquid |
| 1453-58-3 | |
| 204°C | |
| MFCD00005240,MFCD08685900 | |
| 2810 | |
| XKVUYEYANWFIJX-UHFFFAOYSA-N | |
| 5-methyl-1H-pyrazole | |
| 15073 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 1453-58-3 | |
| 82.11 | |
| XKVUYEYANWFIJX-UHFFFAOYSA-N | |
| 15073 | |
| CC1=CC=NN1 |
| C4H6N2 | |
| MFCD00005240,MFCD08685900 | |
| 3-methylpyrazole, 3-methyl-1h-pyrazole, 1h-pyrazole, 3-methyl, 5-methylpyrazole, 3 5-methylpyrazole, pyrazole, 5-methyl, 1h-pyrazole,5-methyl, 1h-pyrazole, 5-methyl, pyrazole, 3-methyl, unii-w7ku1rro6u | |
| 5-methyl-1H-pyrazole |
Safety and Handling
RTECSNumber : UQ7340000
TSCA : No