missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpyrazole 97.0+%, TCI America™

Catalog No. M05645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M05645G 5 g
M056425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M05645G Supplier TCI America Supplier No. M05645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1453-58-3
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005240,MFCD08685900
InChI Key XKVUYEYANWFIJX-UHFFFAOYSA-N
Synonym 3-methylpyrazole, 3-methyl-1h-pyrazole, 1h-pyrazole, 3-methyl, 5-methylpyrazole, 3 5-methylpyrazole, pyrazole, 5-methyl, 1h-pyrazole,5-methyl, 1h-pyrazole, 5-methyl, pyrazole, 3-methyl, unii-w7ku1rro6u
PubChem CID 15073
IUPAC Name 5-methyl-1H-pyrazole
SMILES CC1=CC=NN1

Specifications

Color Yellow
Boiling Point 204°C
Quantity 5 g
UN Number 2810
Formula Weight 82.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylpyrazole
RTECSNumber UQ7340000
TSCA No