missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpyrazole, 99%, Thermo Scientific™

Chemical Identifiers

CAS 1453-58-3
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005240,MFCD08685900
InChI Key XKVUYEYANWFIJX-UHFFFAOYSA-N
Synonym 3-methylpyrazole, 3-methyl-1h-pyrazole, 1h-pyrazole, 3-methyl, 5-methylpyrazole, 3 5-methylpyrazole, pyrazole, 5-methyl, 1h-pyrazole,5-methyl, 1h-pyrazole, 5-methyl, pyrazole, 3-methyl, unii-w7ku1rro6u
PubChem CID 15073
IUPAC Name 5-methyl-1H-pyrazole
SMILES CC1=CC=NN1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC127560050
SDS View Documents
Thermo Scientific Chemicals
127560050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC127560250
SDS View Documents
Thermo Scientific Chemicals
127560250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1453-58-3
82.11
XKVUYEYANWFIJX-UHFFFAOYSA-N
15073
CC1=CC=NN1
C4H6N2
MFCD00005240,MFCD08685900
3-methylpyrazole, 3-methyl-1h-pyrazole, 1h-pyrazole, 3-methyl, 5-methylpyrazole, 3 5-methylpyrazole, pyrazole, 5-methyl, 1h-pyrazole,5-methyl, 1h-pyrazole, 5-methyl, pyrazole, 3-methyl, unii-w7ku1rro6u
5-methyl-1H-pyrazole

Specifications

1453-58-3
100.0
204°C
Authentic
Glass bottle
1.496
5g
1.02
Solubility in water: miscible
CC1=CC=NN1
82.11
82.1
3-Methylpyrazole, 99%
98.5
1.0200g/mL
93°C
98.5% min. (GC)
C4H6N2
MFCD00005240,MFCD08685900
23, 50
3-methylpyrazole, 3-methyl-1h-pyrazole, 1h-pyrazole, 3-methyl, 5-methylpyrazole, 3 5-methylpyrazole, pyrazole, 5-methyl, 1h-pyrazole,5-methyl, 1h-pyrazole, 5-methyl, pyrazole, 3-methyl, unii-w7ku1rro6u
XKVUYEYANWFIJX-UHFFFAOYSA-N
5-methyl-1H-pyrazole
15073
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 215-925-7

RTECSNumber : UQ7340000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only