missing translation for 'onlineSavingsMsg'
Learn More

3-Methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. m13141g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M13141G Supplier TCI America Supplier No. M13141G
Only null left

Chemical Identifiers

CAS 17933-03-8
Molecular Formula C7H9BO2
Molecular Weight (g/mol) 135.96
MDL Number MFCD00040198
InChI Key BJQCPCFFYBKRLM-UHFFFAOYSA-N
Synonym 3-tolylboronic acid, m-tolylboronic acid, 3-methylphenyl boronic acid, 3-methylbenzeneboronic acid, 3-methyl phenyl boronic acid, 3-tolyboronic acid, 3-methylphenyl boranediol, m-methylphenylboronic acid, m-tolyl boronic acid
PubChem CID 2733950
IUPAC Name (3-methylphenyl)boronic acid
SMILES CC1=CC=CC(=C1)B(O)O

Specifications

Quantity 1 g
Formula Weight 135.96
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylphenylboronic Acid (contains varying amounts of Anhydride)
RTECSNumber ED7788888
TSCA No