missing translation for 'onlineSavingsMsg'
Learn More

3'-Methylphenoxyacetamide, TCI America™

Catalog No. M118425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M118425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M118425G Supplier TCI America Supplier No. M118425G

Chemical Identifiers

CAS 10017-53-5
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00191464
InChI Key GOVBTAZVUFGTFB-UHFFFAOYSA-N
PubChem CID 730001
IUPAC Name 2-(3-methylphenoxy)acetamide
SMILES CC1=CC(OCC(N)=O)=CC=C1

Specifications

Melting Point 118°C
Color White
Quantity 25 g
Formula Weight 165.19
Physical Form Crystalline Powder
Chemical Name or Material 3′-Methylphenoxyacetamide
EINECSNumber (3)-4168
TSCA No