missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylnonane 98.0+%, TCI America™
Supplier: TCI America M02825ML
Specifications
| 3-Methylnonane | |
| Colorless | |
| C10H22 | |
| 5 mL | |
| PLZDDPSCZHRBOY-UHFFFAOYNA-N | |
| 3-methylnonane | |
| 22202 | |
| ≥98.0% (GC) |
| 5911-04-6 | |
| 167°C | |
| MFCD00027308 | |
| 3295 | |
| CCCCCCC(C)CC | |
| 142.29 | |
| 142.29 | |
| Liquid |
Chemical Identifiers
| 5911-04-6 | |
| 142.29 | |
| PLZDDPSCZHRBOY-UHFFFAOYNA-N | |
| 3-methylnonane |
| C10H22 | |
| MFCD00027308 | |
| 22202 | |
| CCCCCCC(C)CC |
Safety and Handling
EINECSNumber : (2)-0010
TSCA : No