missing translation for 'onlineSavingsMsg'
Learn More

3-Methylisoxazole-5-carboxylic acid, 97%

Catalog No. p-7055217
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63740MD 250 mg
AAH6374003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63740MD Supplier Thermo Scientific Chemicals Supplier No. H63740MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4857-42-5 | C5H5NO3 | 127.099 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4857-42-5
Molecular Formula C5H5NO3
Molecular Weight (g/mol) 127.099
MDL Number MFCD00464222
InChI Key HXIYCKAAQPHZBM-UHFFFAOYSA-N
Synonym 3-methylisoxazole-5-carboxylic acid, 3-methyl-5-isoxazolecarboxylic acid, 5-isoxazolecarboxylic acid, 3-methyl, 3-methyl-isoxazole-5-carboxylic acid, 5-carboxy-3-methylisoxazole, 5-isoxazolecarboxylicacid, 3-methyl, 3-methylisoxazole-5-carboxylicacid, acmc-1aruu, 3-methyl-5-carboxyisoxazole, aronis24519
PubChem CID 853085
IUPAC Name 3-methyl-1,2-oxazole-5-carboxylic acid
SMILES CC1=NOC(=C1)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 127.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Methylisoxazole-5-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-454-9
RTECSNumber NY2560000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only