missing translation for 'onlineSavingsMsg'
Learn More

3-Methylindole-2-carboxaldehyde, 97%

CAS: 5257-24-9 | C10H9NO | 159.19 g/mol

Supplier:  Thermo Scientific Chemicals H6639303

 View more versions of this product

Catalog No. AAH6639303


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Methylindole-2-carboxaldehyde
C10H9NO
1 g
Air and moisture sensitive
GHSUORIVDBPKKQ-UHFFFAOYSA-N
3-methyl-1H-indole-2-carbaldehyde
277260
97%
5257-24-9
MFCD00957066
4034
3-methylindole-2-carbaldehyde, 3-methyl-1h-indole-2-carboxaldehyde, 3-methylindole-2-carboxaldehyde, 1h-indole-2-carboxaldehyde, 3-methyl, methylindolaldehyd, 3-methyl-2-indolecarbaldehyde, 3-methyl-indole-2-carbaldehyde, 2-formyl-3-methylindole, 3-methyl-2-hydroxymethylenindol, 3-methyl-1h-indole-2-carbaldehyd
CC1=C(NC2=CC=CC=C12)C=O
159.19
159.18
Solid

Chemical Identifiers

5257-24-9
159.19
GHSUORIVDBPKKQ-UHFFFAOYSA-N
277260
CC1=C(NC2=CC=CC=C12)C=O
C10H9NO
MFCD00957066
3-methylindole-2-carbaldehyde, 3-methyl-1h-indole-2-carboxaldehyde, 3-methylindole-2-carboxaldehyde, 1h-indole-2-carboxaldehyde, 3-methyl, methylindolaldehyd, 3-methyl-2-indolecarbaldehyde, 3-methyl-indole-2-carbaldehyde, 2-formyl-3-methylindole, 3-methyl-2-hydroxymethylenindol, 3-methyl-1h-indole-2-carbaldehyd
3-methyl-1H-indole-2-carbaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only