missing translation for 'onlineSavingsMsg'
Learn More

3-Methylglutaric Anhydride 98.0+%, TCI America™

Catalog No. M101010G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M101010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M101010G Supplier TCI America Supplier No. M101010G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4166-53-4
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.127
MDL Number MFCD00006683
InChI Key MGICRVTUCPFQQZ-UHFFFAOYSA-N
Synonym 3-methylglutaric anhydride, 4-methyldihydro-2h-pyran-2,6 3h-dione, 3-methylglutaricanhydride, .beta.-methylglutaric anhydride, acmc-1ago6, 2h-pyran-2,6 3h-dione, dihydro-4-methyl, 3-methyl glutaric anhydride, beta-methylglutaric anhydride, 4-methyltetrahydropyran-2,6-dione
PubChem CID 77828
IUPAC Name 4-methyloxane-2,6-dione
SMILES CC1CC(=O)OC(=O)C1

Specifications

Melting Point 45°C
Color White
Boiling Point 182°C
Quantity 10 g
Formula Weight 128.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylglutaric Anhydride