missing translation for 'onlineSavingsMsg'
Learn More

3-Methylflavone-8-carboxylic Acid 98.0+%, TCI America™

Catalog No. M23715G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23715G 5 g
M237125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23715G Supplier TCI America Supplier No. M23715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3468-01-7
Molecular Formula C17H12O4
Molecular Weight (g/mol) 280.28
MDL Number MFCD00040998
InChI Key KMMBBZOSQNLLMN-UHFFFAOYSA-N
Synonym 3-methylflavone-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl-4h-chromene-8-carboxylic acid, 8-carboxy-3-methylflavone, 4h-1-benzopyran-8-carboxylicacid, 3-methyl-4-oxo-2-phenyl, unii-o8rcm70c48, 4h-1-benzopyran-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl, 3-methyl-4-oxo-2-phenyl-4h-1-benzopyran-8-carboxylic acid, enamine_005545, 3-methylflavone-8-carboxylicacid, acmc-20a6bq
PubChem CID 77016
ChEBI CHEBI:61237
IUPAC Name 3-methyl-4-oxo-2-phenylchromene-8-carboxylic acid
SMILES CC1=C(OC2=C(C1=O)C=CC=C2C(=O)O)C3=CC=CC=C3

Specifications

Melting Point 236°C
Color White-Yellow
Quantity 5 g
Formula Weight 280.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylflavone-8-carboxylic Acid