missing translation for 'onlineSavingsMsg'
Learn More

3-Methylenecyclobutanecarbonitrile, 97%

Catalog No. AC376010250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC376010250 25 g
AC376010050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC376010250 Supplier Thermo Scientific Chemicals Supplier No. 376010250

CAS: 15760-35-7 | C6H7N | 93.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15760-35-7
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
InChI Key ZRWMAMOBIQQJSA-UHFFFAOYSA-N
Synonym 3-methylenecyclobutanecarbonitrile, 3-methylenecyclobutane-1-carbonitrile, cyclobutanecarbonitrile, 3-methylene, 3-methylenecyclobutane carbonitrile, 3-methylenecyanocyclobutane, 3-methylene cyclobutyl carbonitrile, 3-methylene-1-cyanocyclobutane, 1-cyano-3-methylidenecyclobutane, pubchem10218, 3-methylenecyclobutanenitrile
PubChem CID 27474
IUPAC Name 3-methylidenecyclobutane-1-carbonitrile
SMILES C=C1CC(C1)C#N

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula CH2=C4H5CN
Quantity 25 g
Formula Weight 93.13
Percent Purity 97%
Chemical Name or Material 3-Methylenecyclobutanecarbonitrile
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fre

RUO – Research Use Only