missing translation for 'onlineSavingsMsg'
Learn More

3-Methylcyclopentanone, 99%

Catalog No. AAL0494006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0494006 5 g
AAL0494003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0494006 Supplier Thermo Scientific Chemicals Supplier No. L0494006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1757-42-2 | C6H10O | 98.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1757-42-2
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00001416
InChI Key AOKRXIIIYJGNNU-UHFFFAOYSA-N
Synonym 3-methylcyclopentanone, cyclopentanone, 3-methyl, dl-3-methylcyclopentanone, 3-methyl-1-cyclopentanone, cyclopentanone, 3-methyl-, r, 3-methylcyclopentanone, +/-, acmc-20apjx, 3-methyl cyclopentanone, 3-methyl-cyclopentanone, acmc-1ba4k
PubChem CID 15650
IUPAC Name 3-methylcyclopentan-1-one
SMILES CC1CCC(=O)C1

Specifications

Melting Point -58°C
Density 0.914
Boiling Point 144°C to 146°C
Flash Point 36°C (96°F)
Refractive Index 1.434
Quantity 5 g
UN Number UN1224
Beilstein 1305074
Formula Weight 98.15
Percent Purity 99%
Chemical Name or Material 3-Methylcyclopentanone
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 217-148-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only