missing translation for 'onlineSavingsMsg'
Learn More

3-Methylcrotonic Acid 98.0+%, TCI America™

Catalog No. M054325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M054325G 25 g
M0543100G 100 g
M0543500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M054325G Supplier TCI America Supplier No. M054325G

Chemical Identifiers

CAS 541-47-9
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00004366
InChI Key YYPNJNDODFVZLE-UHFFFAOYSA-N
Synonym 3,3-dimethylacrylic acid, senecioic acid, 3-methylcrotonic acid, 3-methyl-2-butenoic acid, 2-butenoic acid, 3-methyl, senecic acid, beta,beta-dimethylacrylic acid, crotonic acid, 3-methyl, beta,beta-dimethacrylic acid, beta-methylcrotonic acid
PubChem CID 10931
ChEBI CHEBI:37127
IUPAC Name 3-methylbut-2-enoic acid
SMILES CC(=CC(=O)O)C

Specifications

Melting Point 68°C
Color White-Yellow
Boiling Point 98°C
Quantity 25 g
UN Number 3261
Formula Weight 100.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylcrotonic Acid
EINECSNumber (2)-0609
RTECSNumber GQ5425000
TSCA Yes