missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbutanamide, 97%, Thermo Scientific™

$88.44 - $88.44

Chemical Identifiers

CAS 541-46-8
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00014807
InChI Key SANOUVWGPVYVAV-UHFFFAOYSA-N
Synonym isovaleramide, butanamide, 3-methyl, 3-methylbutyramide, isovaleric amide, isopentanamide, beta-methylbutyramide, isovaleric acid amide, unii-9cp4kb634m, isovaleramide usan, .beta.-methylbutyramide
PubChem CID 10930
IUPAC Name 3-methylbutanamide
SMILES CC(C)CC(=O)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BTB12988EA
SDS View Documents
Thermo Scientific Chemicals
BTB12988EA
10 g
Each for $88.44
Only null left
 
Description

Description

Specifications

Chemical Identifiers

541-46-8
101.149
SANOUVWGPVYVAV-UHFFFAOYSA-N
10930
CC(C)CC(=O)N
C5H11NO
MFCD00014807
isovaleramide, butanamide, 3-methyl, 3-methylbutyramide, isovaleric amide, isopentanamide, beta-methylbutyramide, isovaleric acid amide, unii-9cp4kb634m, isovaleramide usan, .beta.-methylbutyramide
3-methylbutanamide

Specifications

541-46-8
100.0
C5H11NO
10 g
SANOUVWGPVYVAV-UHFFFAOYSA-N
3-methylbutanamide
10930
97%
96.5
97%
MFCD00014807
isovaleramide, butanamide, 3-methyl, 3-methylbutyramide, isovaleric amide, isopentanamide, beta-methylbutyramide, isovaleric acid amide, unii-9cp4kb634m, isovaleramide usan, .beta.-methylbutyramide
CC(C)CC(=O)N
101.149
101.15
3-Methylbutanamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents