missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzo[b]thiophene-2-carboxaldehyde, 98%

CAS: 22053-74-3 | C10H8OS | 176.233 g/mol

$156.75 - $577.26

Chemical Identifiers

CAS 22053-74-3
Molecular Formula C10H8OS
Molecular Weight (g/mol) 176.233
MDL Number MFCD00052307
InChI Key DRZGHNXLEQHVHB-UHFFFAOYSA-N
Synonym 3-methylbenzo b thiophene-2-carboxaldehyde, 3-methylbenzo b thiophene-2-carbaldehyde, 3-methyl-benzo b thiophene-2-carbaldehyde, pubchem7453, acmc-20an0e, 3-methylthianaphthene-2-carboxaldehyde, 3-methyl-2-benzothiophenecarboxaldehyde, 3-methyl benzo b thiophene-2-carbaldehyde, 3-methylbenzo b thiophene-2 carboxaldehyde
PubChem CID 519918
IUPAC Name 3-methyl-1-benzothiophene-2-carbaldehyde
SMILES CC1=C(SC2=CC=CC=C12)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1137503
View Documents
Thermo Scientific Chemicals
A1137503
1 g
Each for $156.75
Only null left
 
AAA1137506
View Documents
Thermo Scientific Chemicals
A1137506
5 g
Each for $577.26
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

22053-74-3
176.233
DRZGHNXLEQHVHB-UHFFFAOYSA-N
519918
CC1=C(SC2=CC=CC=C12)C=O
C10H8OS
MFCD00052307
3-methylbenzo b thiophene-2-carboxaldehyde, 3-methylbenzo b thiophene-2-carbaldehyde, 3-methyl-benzo b thiophene-2-carbaldehyde, pubchem7453, acmc-20an0e, 3-methylthianaphthene-2-carboxaldehyde, 3-methyl-2-benzothiophenecarboxaldehyde, 3-methyl benzo b thiophene-2-carbaldehyde, 3-methylbenzo b thiophene-2 carboxaldehyde
3-methyl-1-benzothiophene-2-carbaldehyde

Specifications

22053-74-3
C10H8OS
1 g
Air Sensitive
DRZGHNXLEQHVHB-UHFFFAOYSA-N
3-methyl-1-benzothiophene-2-carbaldehyde
519918
98%
88°C to 90°C
MFCD00052307
121582
3-methylbenzo b thiophene-2-carboxaldehyde, 3-methylbenzo b thiophene-2-carbaldehyde, 3-methyl-benzo b thiophene-2-carbaldehyde, pubchem7453, acmc-20an0e, 3-methylthianaphthene-2-carboxaldehyde, 3-methyl-2-benzothiophenecarboxaldehyde, 3-methyl benzo b thiophene-2-carbaldehyde, 3-methylbenzo b thiophene-2 carboxaldehyde
CC1=C(SC2=CC=CC=C12)C=O
176.233
176.24
3-Methylbenzo[b]thiophene-2-carboxaldehyde
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only