missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzo[b]thiophene-2-carboxaldehyde, 98%

Catalog No. AAA1137503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1137503 1 g
AAA1137506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1137503 Supplier Thermo Scientific Chemicals Supplier No. A1137503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22053-74-3 | C10H8OS | 176.233 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22053-74-3
Molecular Formula C10H8OS
Molecular Weight (g/mol) 176.233
MDL Number MFCD00052307
InChI Key DRZGHNXLEQHVHB-UHFFFAOYSA-N
Synonym 3-methylbenzo b thiophene-2-carboxaldehyde, 3-methylbenzo b thiophene-2-carbaldehyde, 3-methyl-benzo b thiophene-2-carbaldehyde, pubchem7453, acmc-20an0e, 3-methylthianaphthene-2-carboxaldehyde, 3-methyl-2-benzothiophenecarboxaldehyde, 3-methyl benzo b thiophene-2-carbaldehyde, 3-methylbenzo b thiophene-2 carboxaldehyde
PubChem CID 519918
IUPAC Name 3-methyl-1-benzothiophene-2-carbaldehyde
SMILES CC1=C(SC2=CC=CC=C12)C=O

Specifications

Melting Point 88°C to 90°C
Quantity 1 g
Beilstein 121582
Sensitivity Air Sensitive
Formula Weight 176.24
Percent Purity 98%
Chemical Name or Material 3-Methylbenzo[b]thiophene-2-carboxaldehyde
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only