missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylaminomethyl)pyridine 98.0+%, TCI America™

Catalog No. M22571G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22571G 1 g
M22575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22571G Supplier TCI America Supplier No. M22571G
Only null left

Chemical Identifiers

CAS 20173-04-0
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00023610
InChI Key MCSAQVGDZLPTBS-UHFFFAOYSA-N
Synonym n-methyl-1-pyridin-3-yl methanamine, n-methyl-n-3-pyridylmethyl amine, 3-methylaminomethyl pyridine, n-methyl pyridin-3-yl methanamine, 3-methylamino methyl pyridine, 3-pyridinemethanamine, n-methyl, 3-picolylmethylamine, n-methyl-3-pyridylmethylamine, methyl pyridin-3-ylmethyl amine, n-methyl-3-pyridinemethylamine
PubChem CID 88393
IUPAC Name N-methyl-1-pyridin-3-ylmethanamine
SMILES CNCC1=CN=CC=C1

Specifications

Color Yellow
Boiling Point 98°C
Quantity 1 g
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Methylaminomethyl)pyridine