missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylamino)propionitrile 98.0+%, TCI America™

Catalog No. M231225ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M231225ML 25 mL
M2312100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M231225ML Supplier TCI America Supplier No. M231225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 693-05-0
Molecular Formula C4H8N2
Molecular Weight (g/mol) 84.12
MDL Number MFCD00001954
InChI Key UNIJBMUBHBAUET-UHFFFAOYSA-N
Synonym 3-methylamino propanenitrile, 3-methylaminopropionitrile, 3-methylamino propionitrile, propanenitrile, 3-methylamino, 2-cyanoethyl methyl amine, propionitrile, 3-methylamino, n-2-cyanoethyl methylamine, 3-n-methylamino propionitrile, 2-cyanoethyl methylamine, n-methyl-beta-alaninenitrile
PubChem CID 69656
IUPAC Name 3-(methylamino)propanenitrile
SMILES CNCCC#N

Specifications

Color Yellow
Boiling Point 81°C
Quantity 25 mL
Formula Weight 84.12
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Methylamino)propionitrile
RTECSNumber TZ4910000
TSCA Yes