missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylamino)-1-propanol 97.0+%, TCI America™

Catalog No. p-7132758
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
M14845G 5 g
M148425G 25 g
2 options

Catalog No. M14845G

Supplier: TCI America M14845G

Only null left

Chemical Identifiers

CAS 42055-15-2
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
InChI Key KRGXWTOLFOPIKV-UHFFFAOYSA-N
Synonym 3-methylamino propan-1-ol, 3-methylamino-1-propanol, 3-methylaminopropanol, 3-methylamino-propanol, 3-methylamino propanol, acmc-1argo, 3-methylamino-propan-1-ol, 3-hydroxypropyl methylamine, n-methyl-3-hydroxypropylamine
PubChem CID 10148986
IUPAC Name 3-(methylamino)propan-1-ol
SMILES CNCCCO

Specifications

Color Yellow
Boiling Point 169°C
Quantity 5 g
UN Number 2735
Formula Weight 89.14
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Methylamino)-1-propanol