missing translation for 'onlineSavingsMsg'
Learn More

3-Methylamino-1-propanol, 95%

Catalog No. AAH5881806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5881806 5 g
AAH5881803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5881806 Supplier Thermo Scientific Chemicals Supplier No. H5881806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 42055-15-2 | C4H11NO | 89.138 g/mol

3-Methylamino-1-propanol is an alkanolamine that is used to redisperse compacted solids (such as antiperspirant materials) in order to analyze its components.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylamino-1-propanol is an alkanolamine that is used to redisperse compacted solids (such as antiperspirant materials) in order to analyze its components.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 42055-15-2
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD06797456
InChI Key KRGXWTOLFOPIKV-UHFFFAOYSA-N
Synonym 3-methylamino propan-1-ol, 3-methylamino-1-propanol, 3-methylaminopropanol, 3-methylamino-propanol, 3-methylamino propanol, acmc-1argo, 3-methylamino-propan-1-ol, 3-hydroxypropyl methylamine, n-methyl-3-hydroxypropylamine
PubChem CID 10148986
IUPAC Name 3-(methylamino)propan-1-ol
SMILES CNCCCO

Specifications

Density 0.98
Boiling Point 169°C
Flash Point 82°C (180°F)
Refractive Index 1.44
Linear Formula CH3NH(CH2)3OH
Quantity 5 g
UN Number UN2735
Solubility Information Slightly soluble in water.
Formula Weight 89.14
Percent Purity 95%
Chemical Name or Material 3-Methylamino-1-propanol
Show More Show Less
Hazard Category H227-H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only