missing translation for 'onlineSavingsMsg'
Learn More

3'-Methylacetophenone 97.0+%, TCI America™

Catalog No. M019125ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M019125ML 25 mL
M0191100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M019125ML Supplier TCI America Supplier No. M019125ML

Chemical Identifiers

CAS 585-74-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008742
InChI Key FSPSELPMWGWDRY-UHFFFAOYSA-N
Synonym 3'-methylacetophenone, 1-m-tolyl ethanone, 3-methylacetophenone, m-methylacetophenone, ethanone, 1-3-methylphenyl, acetophenone, m-methyl, methyl m-tolyl ketone, 1-3-methylphenyl ethan-1-one, 3-acetyltoluene, acetophenone, 3'-methyl
PubChem CID 11455
IUPAC Name 1-(3-methylphenyl)ethan-1-one
SMILES CC(=O)C1=CC=CC(C)=C1

Specifications

Color Yellow
Boiling Point 219°C
Quantity 25 mL
Formula Weight 134.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3′-Methylacetophenone
EINECSNumber (3)-1239
TSCA No