missing translation for 'onlineSavingsMsg'
Learn More

3'-Methylacetophenone, 97%

CAS: 585-74-0 | C9H10O | 134.18 g/mol

Supplier:  Thermo Scientific Chemicals L0044014

 View more versions of this product

Catalog No. AAL0044014


Only null left
Explore available promotions

Description

Description

3'-Methylacetophenone is used in Pharmaceutical Intermediates and in Organic Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′-Methylacetophenone is used in Pharmaceutical Intermediates and in Organic Intermediates.

Solubility
Difficult to mix.

Notes
Protect from heat. Store away from oxidizing agents.
Specifications

Specifications

3'-Methylacetophenone
-9°C
219°C
C9H10O
MFCD00008742
956637
Difficult to mix.
CC(=O)C1=CC=CC(C)=C1
134.18
134.18
585-74-0
1.01
85°C (185°F)
1.531
25 g
3'-methylacetophenone, 1-m-tolyl ethanone, 3-methylacetophenone, m-methylacetophenone, ethanone, 1-3-methylphenyl, acetophenone, m-methyl, methyl m-tolyl ketone, 1-3-methylphenyl ethan-1-one, 3-acetyltoluene, acetophenone, 3'-methyl
FSPSELPMWGWDRY-UHFFFAOYSA-N
1-(3-methylphenyl)ethanone
11455
97%

Chemical Identifiers

585-74-0
134.18
FSPSELPMWGWDRY-UHFFFAOYSA-N
11455
CC(=O)C1=CC=CC(C)=C1
C9H10O
MFCD00008742
3'-methylacetophenone, 1-m-tolyl ethanone, 3-methylacetophenone, m-methylacetophenone, ethanone, 1-3-methylphenyl, acetophenone, m-methyl, methyl m-tolyl ketone, 1-3-methylphenyl ethan-1-one, 3-acetyltoluene, acetophenone, 3'-methyl
1-(3-methylphenyl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 209-561-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only