missing translation for 'onlineSavingsMsg'
Learn More

3'-Methylacetophenone, 97%

Catalog No. AAL0044006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0044006 5 g
AAL0044014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0044006 Supplier Thermo Scientific Chemicals Supplier No. L0044006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 585-74-0 | C9H10O | 134.18 g/mol

3'-Methylacetophenone is used in Pharmaceutical Intermediates and in Organic Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′-Methylacetophenone is used in Pharmaceutical Intermediates and in Organic Intermediates.

Solubility
Difficult to mix.

Notes
Protect from heat. Store away from oxidizing agents.

Chemical Identifiers

CAS 585-74-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008742
InChI Key FSPSELPMWGWDRY-UHFFFAOYSA-N
Synonym 3'-methylacetophenone, 1-m-tolyl ethanone, 3-methylacetophenone, m-methylacetophenone, ethanone, 1-3-methylphenyl, acetophenone, m-methyl, methyl m-tolyl ketone, 1-3-methylphenyl ethan-1-one, 3-acetyltoluene, acetophenone, 3'-methyl
PubChem CID 11455
IUPAC Name 1-(3-methylphenyl)ethanone
SMILES CC(=O)C1=CC=CC(C)=C1

Specifications

Melting Point -9°C
Density 1.01
Boiling Point 219°C
Flash Point 85°C (185°F)
Refractive Index 1.531
Quantity 5 g
Beilstein 956637
Solubility Information Difficult to mix.
Formula Weight 134.18
Percent Purity 97%
Chemical Name or Material 3'-Methylacetophenone
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 209-561-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only