missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-L-phenylalanine, 95%

Catalog No. AAH52047MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52047MD 250 mg
AAH5204703 1 g
AAH5204706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52047MD Supplier Thermo Scientific Chemicals Supplier No. H52047MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 114926-37-3 | C10H13NO2 | 179.219 g/mol

It finds its application in the identification of N-acyl 4-(5-pyrimidine-2,4-dionyl)phenylalanine derivatives and their orally active prodrug esters as dual-acting alpha4-beta1 and alpha4-beta7 receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the identification of N-acyl 4-(5-pyrimidine-2,4-dionyl)phenylalanine derivatives and their orally active prodrug esters as dual-acting alpha4-beta1 and alpha4-beta7 receptor antagonists.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 114926-37-3
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00044972
InChI Key JZRBSTONIYRNRI-VIFPVBQESA-N
Synonym 3-methyl-l-phenylalanine, l-3-methylphenylalanine, 3-methylphenyl-l-alanine, 3-methy-l-phenylalanine, s-2-amino-3-m-tolyl propanoic acid, 3-methylphenylalanine, l-3-methylphe, phenylalanine, 3-methyl, 2s-2-amino-3-3-methylphenyl propanoic acid, l-3-me-phe-oh
PubChem CID 2761494
IUPAC Name (2S)-2-amino-3-(3-methylphenyl)propanoic acid
SMILES CC1=CC(=CC=C1)CC(C(=O)O)N

Specifications

Solubility Information Slightly soluble in water.
Quantity 250 mg
Formula Weight 179.22
Percent Purity 95%
Chemical Name or Material 3-Methyl-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only