missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-L-phenylalanine, 95%

CAS: 114926-37-3 | C10H13NO2 | 179.219 g/mol

Supplier:  Thermo Scientific Chemicals H5204703

Quantity 1 g

 View more versions of this product

Catalog No. AAH5204703


Only null left

Description

Description

It finds its application in the identification of N-acyl 4-(5-pyrimidine-2,4-dionyl)phenylalanine derivatives and their orally active prodrug esters as dual-acting alpha4-beta1 and alpha4-beta7 receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the identification of N-acyl 4-(5-pyrimidine-2,4-dionyl)phenylalanine derivatives and their orally active prodrug esters as dual-acting alpha4-beta1 and alpha4-beta7 receptor antagonists.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

114926-37-3
179.219
JZRBSTONIYRNRI-VIFPVBQESA-N
2761494
CC1=CC(=CC=C1)CC(C(=O)O)N
C10H13NO2
MFCD00044972
3-methyl-l-phenylalanine, l-3-methylphenylalanine, 3-methylphenyl-l-alanine, 3-methy-l-phenylalanine, s-2-amino-3-m-tolyl propanoic acid, 3-methylphenylalanine, l-3-methylphe, phenylalanine, 3-methyl, 2s-2-amino-3-3-methylphenyl propanoic acid, l-3-me-phe-oh
(2S)-2-amino-3-(3-methylphenyl)propanoic acid

Specifications

3-Methyl-L-phenylalanine
C10H13NO2
3-methyl-l-phenylalanine, l-3-methylphenylalanine, 3-methylphenyl-l-alanine, 3-methy-l-phenylalanine, s-2-amino-3-m-tolyl propanoic acid, 3-methylphenylalanine, l-3-methylphe, phenylalanine, 3-methyl, 2s-2-amino-3-3-methylphenyl propanoic acid, l-3-me-phe-oh
JZRBSTONIYRNRI-VIFPVBQESA-N
(2S)-2-amino-3-(3-methylphenyl)propanoic acid
179.219
179.22
114926-37-3
MFCD00044972
Slightly soluble in water.
CC1=CC(=CC=C1)CC(C(=O)O)N
1 g
2761494
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions