missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-6-nitroindazole 98.0+%, TCI America™
$55.92 - $160.30
Chemical Identifiers
| CAS | 6494-19-5 |
|---|---|
| Molecular Formula | C8H7N3O2 |
| Molecular Weight (g/mol) | 177.163 |
| MDL Number | MFCD07082704 |
| InChI Key | FUNWSYKLFDLUIZ-UHFFFAOYSA-N |
| Synonym | 3-methyl-6-nitro-1h-indazole, 3-methyl-6-nitroindazole, 1h-indazole, 3-methyl-6-nitro, 3-methyl-6-nitro-indazole, pubchem9492, pubchem18398, 6-nitro-3-methylindazole, ksc352s0d, 2h-indazole,3-methyl-6-nitro |
| PubChem CID | 10921054 |
| IUPAC Name | 3-methyl-6-nitro-2H-indazole |
| SMILES | CC1=C2C=CC(=CC2=NN1)[N+](=O)[O-] |
Chemical Identifiers
| 6494-19-5 | |
| 177.163 | |
| FUNWSYKLFDLUIZ-UHFFFAOYSA-N | |
| 10921054 | |
| CC1=C2C=CC(=CC2=NN1)[N+](=O)[O-] |
| C8H7N3O2 | |
| MFCD07082704 | |
| 3-methyl-6-nitro-1h-indazole, 3-methyl-6-nitroindazole, 1h-indazole, 3-methyl-6-nitro, 3-methyl-6-nitro-indazole, pubchem9492, pubchem18398, 6-nitro-3-methylindazole, ksc352s0d, 2h-indazole,3-methyl-6-nitro | |
| 3-methyl-6-nitro-2H-indazole |
Specifications
| 6494-19-5 | |
| C8H7N3O2 | |
| 5 g | |
| FUNWSYKLFDLUIZ-UHFFFAOYSA-N | |
| 3-methyl-6-nitro-2H-indazole | |
| 10921054 | |
| ≥98.0% (GC) | |
| 3-Methyl-6-nitroindazole |
| 186°C | |
| MFCD07082704 | |
| 3-methyl-6-nitro-1h-indazole, 3-methyl-6-nitroindazole, 1h-indazole, 3-methyl-6-nitro, 3-methyl-6-nitro-indazole, pubchem9492, pubchem18398, 6-nitro-3-methylindazole, ksc352s0d, 2h-indazole,3-methyl-6-nitro | |
| CC1=C2C=CC(=CC2=NN1)[N+](=O)[O-] | |
| 177.163 | |
| 177.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No