missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-pyrazolone 98.0+%, TCI America™

Catalog No. p-7132200
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M049125G 25 g
M0491500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M049125G Supplier TCI America Supplier No. M049125G

Chemical Identifiers

CAS 108-26-9
Molecular Formula C4H6N2O
Molecular Weight (g/mol) 98.105
MDL Number MFCD00020699
InChI Key NHLAPJMCARJFOG-UHFFFAOYSA-N
Synonym 3-methyl-2-pyrazolin-5-one, 3-methyl-5-pyrazolone, 3-methyl-1h-pyrazol-5 4h-one, 3h-pyrazol-3-one, 2,4-dihydro-5-methyl, 3-methyl-pyrazolon-5, 2-pyrazolin-5-one, 3-methyl, 3-methyl-4,5-dihydro-1h-pyrazol-5-one, 5-methyl-2,4-dihydro-3h-pyrazol-3-one, 2,4-dihydro-5-methyl-3h-pyrazol-3-one, 3-methyl-pyrazolon-5 german
PubChem CID 7920
IUPAC Name 3-methyl-1,4-dihydropyrazol-5-one
SMILES CC1=NNC(=O)C1

Specifications

Melting Point 222°C
Color White-Yellow
Quantity 25 g
Formula Weight 98.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-5-pyrazolone
EINECSNumber (5)-0284
RTECSNumber UQ9451500
TSCA Yes