missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-phenyl-4-isoxazolecarboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC20301EA

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC20301EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

17153-21-8
203.197
3-methyl-5-phenylisoxazole-4-carboxylic acid, 3-methyl-5-phenyl-4-isoxazolecarboxylic acid, 4-carboxy-3-methylisoxazol-5-yl benzene, 3-methyl-5-phenyl-4-isoxazole carboxylic acid, 3-methyl-5-phenyl-isoxazole-4-carboxylic acid, 4-isoxazolecarboxylicacid, 3-methyl-5-phenyl, 4-isoxazolecarboxylic acid, 3-methyl-5-phenyl
3-methyl-5-phenyl-1,2-oxazole-4-carboxylic acid
C11H9NO3
GLNQCTGGLIXRRJ-UHFFFAOYSA-N
2776514
CC1=NOC(=C1C(=O)O)C2=CC=CC=C2

Specifications

3-Methyl-5-phenyl-4-isoxazolecarboxylic acid
97%
C11H9NO3
3-methyl-5-phenylisoxazole-4-carboxylic acid, 3-methyl-5-phenyl-4-isoxazolecarboxylic acid, 4-carboxy-3-methylisoxazol-5-yl benzene, 3-methyl-5-phenyl-4-isoxazole carboxylic acid, 3-methyl-5-phenyl-isoxazole-4-carboxylic acid, 4-isoxazolecarboxylicacid, 3-methyl-5-phenyl, 4-isoxazolecarboxylic acid, 3-methyl-5-phenyl
CC1=NOC(=C1C(=O)O)C2=CC=CC=C2
203.197
203.2
17153-21-8
Amber Glass Bottle
10g
GLNQCTGGLIXRRJ-UHFFFAOYSA-N
3-methyl-5-phenyl-1,2-oxazole-4-carboxylic acid
2776514
97%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents