missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-phenyl-4-isoxazolecarbaldehyde, 95%, Thermo Scientific™

Catalog No. CC20304DA
Change view
Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC20304DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC20304DA Supplier Thermo Scientific Chemicals Supplier No. CC20304DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89479-66-3
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.198
MDL Number MFCD03086122
InChI Key IZULDLZFUWWFHF-UHFFFAOYSA-N
Synonym 3-methyl-5-phenyl-4-isoxazolecarbaldehyde, 3-methyl-5-phenylisoxazole-4-carbaldehyde, 3-methyl-5-phenyl-isoxazole-4-carbaldehyde, 4-isoxazolecarboxaldehyde,3-methyl-5-phenyl, acmc-20e4b4, 4-formyl-3-methyl-5-phenylisoxazole, 3-methyl-5-phenyl-4-isoxazolecarboxaldehyde
PubChem CID 2776516
IUPAC Name 3-methyl-5-phenyl-1,2-oxazole-4-carbaldehyde
SMILES CC1=NOC(=C1C=O)C2=CC=CC=C2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 187.2
Percent Purity 95%
Chemical Name or Material 3-Methyl-5-phenyl-4-isoxazolecarbaldehyde