missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-phenyl-1H-pyrazole, Thermo Scientific Chemicals

Catalog No. AAH5065003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5065003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5065003 Supplier Thermo Scientific Chemicals Supplier No. H5065003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3347-62-4 | Molecular Formula: C10H10N2 | Molecular Weight: 158.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3347-62-4
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
MDL Number MFCD00022385,MFCD00524752
InChI Key QHRSESMSOJZMCO-UHFFFAOYSA-N
PubChem CID 18774
IUPAC Name 5-methyl-3-phenyl-1H-pyrazole
SMILES CC1=CC(=NN1)C1=CC=CC=C1

Specifications

Melting Point 124°C to 127°C
Quantity 1 g
Synonym 3-methyl-5-phenyl-1h-pyrazole, femerazol, 3-methyl-5-phenylpyrazole, phemerazole, 1h-pyrazole, 3-methyl-5-phenyl, 5-methyl-3-phenylpyrazole, 1h-pyrazole,3-methyl-5-phenyl, pyrazole, 3-methyl-5-phenyl, unii-0xfx19d77a, 1h-pyrazole, 5-methyl-3-phenyl
Formula Weight 158.2
Physical Form Powder
Chemical Name or Material 3-Methyl-5-phenyl-1H-pyrazole
EINECSNumber 222-099-1
RTECSNumber UQ7525000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only