missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-cyclohexene-1,2-dicarboxylic Anhydride 95.0+%, TCI America™

Catalog No. p-7132586
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11901G 1 g
M11905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11901G Supplier TCI America Supplier No. M11901G
Only null left

Chemical Identifiers

CAS 5333-84-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00014585
InChI Key XPEKVUUBSDFMDR-UHFFFAOYNA-N
Synonym 3-Methyl-DELTA4-tetrahydrophthalic Anhydride
PubChem CID 95332
IUPAC Name 4-methyl-1,3,3a,4,7,7a-hexahydro-2-benzofuran-1,3-dione
SMILES CC1C=CCC2C1C(=O)OC2=O

Specifications

Melting Point 62°C
Color White
Quantity 1 g
Formula Weight 166.18
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-4-cyclohexene-1,2-dicarboxylic Anhydride
EINECSNumber (3)-2451&(3)-2479
TSCA Yes