missing translation for 'onlineSavingsMsg'
Learn More

Petroleum ether, pure, boiling range ca. 180-280°C

Catalog No. AC387490050
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Glass Bottle
Metal drum
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC387490050 5 L Plastic drum
AC387490010 1 L Glass Bottle
AC387490025 2.5 L Glass Bottle
AC387490250 25 L Metal drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC387490050 Supplier Thermo Scientific Chemicals Supplier No. 387490050

CAS: 8008-20-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 8008-20-6
MDL Number MFCD00135561

Specifications

Color Colorless
Density 0.7900g/mL
Boiling Point 200.0°C to 250.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Packaging Plastic drum
Refractive Index 1.4300 to 1.4500
Quantity 5 L
Merck Index 15,5342
Specific Gravity 0.79
Solubility Information Solubility in water: insoluble
Grade Pure
Physical Form Liquid
Chemical Name or Material Petroleum ether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
EINECSNumber 232-366-4

RUO – Research Use Only