missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-penten-2-one 95.0+%, TCI America™

Catalog No. M1139500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1139500ML 500 mL
M113925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1139500ML Supplier TCI America Supplier No. M1139500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 565-62-8
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00151839
InChI Key ZAMCMCQRTZKGDX-SNAWJCMRSA-N
Synonym 3-methyl-3-penten-2-one, e-3-methylpent-3-en-2-one, e-3-methyl-3-penten-2-one, unii-jbr4o01db2, jbr4o01db2, 3-methyl-2-penten-4-one, 3-penten-2-one, 3-methyl, 3-methyl-3-pentene-2-one, 2-acetyl-2-butene, 3-methyl-4-oxopent-2-ene
PubChem CID 5364579
IUPAC Name (E)-3-methylpent-3-en-2-one
SMILES CC=C(C)C(=O)C

Specifications

Color Yellow
Boiling Point 140°C
Quantity 500 mL
UN Number 1224
Formula Weight 98.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-penten-2-one