missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-(p-toluenesulfonyloxymethyl)oxetane, 98%

Catalog No. p-7055433
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6401103 1 g
AAH6401106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6401103 Supplier Thermo Scientific Chemicals Supplier No. H6401103

CAS: 99314-44-0 | C12H16O4S | 256.316 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99314-44-0
Molecular Formula C12H16O4S
Molecular Weight (g/mol) 256.316
MDL Number MFCD08275394
InChI Key PZOQQSOZRVZCMC-UHFFFAOYSA-N
Synonym 3-methyloxetan-3-yl methyl 4-methylbenzenesulfonate, 3-methyl-3-oxetanyl methyl 4-methylbenzenesulfonate, 3-methyloxetan-3-yl methyl p-tosylate, 3-methyl-3-toluenesulfonyloxymethyl oxetane, 3-methyloxetan-3-yl methyl 4-methylbenzene-1-sulfonate, upcmld00wcrh3-139, 3-tosyloxymethyl-3-methyloxetane, 3-methyl-3-oxetanyl methyl p-tosylate, methyloxetanylmethylmethylbenzenesulfonate, 3-methyloxetan-3-yl methyl 4-toluenesulfonate
PubChem CID 5461213
IUPAC Name (3-methyloxetan-3-yl)methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2(COC2)C

Specifications

Melting Point 50°C to 51°C
Color White
Quantity 1 g
Beilstein 4441465
Formula Weight 256.32
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Methyl-3-(p-toluenesulfonyloxymethyl)oxetane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only