missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-oxetanemethanol, 98%

Catalog No. AC240840100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC240840100 10 g Glass bottle
AC240840500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC240840100 Supplier Thermo Scientific Chemicals Supplier No. 240840100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3143-02-0 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3143-02-0
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00010273
InChI Key NLQMSBJFLQPLIJ-UHFFFAOYSA-N
Synonym 3-methyl-3-oxetanemethanol, 3-methyloxetan-3-yl methanol, 3-hydroxymethyl-3-methyloxetane, 3-oxetanemethanol, 3-methyl, 3-methyl-3-oxetanyl methanol, 3-methyloxetane-3-methanol, 3-methyl-3-oxethanemethanol, 3-methyl-3-hydroxymethyloxetane, 3-hydroxymethyl-3-methyl-oxetane, 3-methyl-oxetan-3-yl-methanol
PubChem CID 137837
IUPAC Name (3-methyloxetan-3-yl)methanol
SMILES CC1(COC1)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0340g/mL
Boiling Point 122.0°C (70.0 mmHg)
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Specific Gravity 1.034
Formula Weight 102.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-oxetanemethanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If s

RUO – Research Use Only