missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-oxetanecarboxylic Acid 98.0+%, TCI America™

Catalog No. M29621G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M29621G Supplier TCI America Supplier No. M29621G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 28562-68-7
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD09842435
InChI Key PMPZEEUNGQSAIQ-UHFFFAOYSA-N
Synonym 3-methyloxetane-3-carboxylicacid, 3-methyl-3-oxetanecarboxylic acid, 3-oxetanecarboxylic acid, 3-methyl, 3-methyl-oxetane-3-carboxylic acid, pubchem14214, ksc199o1l, 3-methyl oxetane-3-carboxylic acid, 3-methyl-3-oxetane carboxylic acid, 3-oxetanecarboxylicacid, 3-methyl, 3-methyloxetane-3-carboxylic acid
PubChem CID 14028997
IUPAC Name 3-methyloxetane-3-carboxylic acid
SMILES CC1(COC1)C(=O)O

Specifications

Melting Point 60°C
Quantity 1 g
Formula Weight 116.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-3-oxetanecarboxylic Acid
TSCA No
Recommended Storage Refrigerator