missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-hexanol, 99%

Catalog No. aal0462514
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
AAL0462514 25 g
AAL0462506 5 g
2 options

Catalog No. AAL0462514

Supplier: Thermo Scientific Chemicals L0462514

Only null left

CAS: 597-96-6 | C7H16O | 116.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 597-96-6
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.204
MDL Number MFCD00021839
InChI Key KYWJZCSJMOILIZ-UHFFFAOYSA-N
Synonym 3-methyl-3-hexanol, 3-hexanol, 3-methyl, 2-ethyl-2-pentanol, 3-methyl-hexanol-3, 3-methyl-hexanol-3 german, acmc-20an0r, 4-01-00-01749 beilstein handbook reference
PubChem CID 11708
IUPAC Name 3-methylhexan-3-ol
SMILES CCCC(C)(CC)O

Specifications

Density 0.823
Boiling Point 143°C
Flash Point 45°C (113°F)
Refractive Index 1.4235
Quantity 25 g
UN Number UN1987
Beilstein 1719032
Formula Weight 116.2
Percent Purity 99%
Chemical Name or Material 3-Methyl-3-hexanol
Hazard Category H226-H302
Hazard Statement GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 209-910-4
RTECSNumber MP0950000
TSCA Yes

RUO – Research Use Only