missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-hexanol, 99%

Catalog No. AAL0462506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0462506 5 g
AAL0462514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0462506 Supplier Thermo Scientific Chemicals Supplier No. L0462506

CAS: 597-96-6 | C7H16O | 116.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 597-96-6
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.204
MDL Number MFCD00021839
InChI Key KYWJZCSJMOILIZ-UHFFFAOYSA-N
Synonym 3-methyl-3-hexanol, 3-hexanol, 3-methyl, 2-ethyl-2-pentanol, 3-methyl-hexanol-3, 3-methyl-hexanol-3 german, acmc-20an0r, 4-01-00-01749 beilstein handbook reference
PubChem CID 11708
IUPAC Name 3-methylhexan-3-ol
SMILES CCCC(C)(CC)O

Specifications

Density 0.823
Boiling Point 143°C
Flash Point 45°C (113°F)
Refractive Index 1.4235
Quantity 5 g
UN Number UN1987
Beilstein 1719032
Formula Weight 116.2
Percent Purity 99%
Chemical Name or Material 3-Methyl-3-hexanol
Hazard Category H226-H302
Hazard Statement GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 209-910-4
RTECSNumber MP0950000
TSCA Yes

RUO – Research Use Only