missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-buten-2-one (stabilized with HQ) 95.0+%, TCI America™

Catalog No. M037825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M037825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M037825ML Supplier TCI America Supplier No. M037825ML
Only null left

Chemical Identifiers

CAS 814-78-8
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00059207
InChI Key ZGHFDIIVVIFNPS-UHFFFAOYSA-N
Synonym Isopropenyl Methyl Ketone
PubChem CID 13143
IUPAC Name 3-methylbut-3-en-2-one
SMILES CC(=C)C(=O)C

Specifications

Color Yellow
Boiling Point 100°C
Quantity 25 mL
UN Number 1246
Formula Weight 84.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-buten-2-one (stabilized with HQ)
EINECSNumber (2)-0564
RTECSNumber EN0175000
TSCA Yes
Recommended Storage Freezer